Learn More
Hach Company Glass pH probe GP Refil. Kit
Catalog No. 5000207315 Shop All Hach Company Products
Encompass
IntelliCAL PHC705 Laboratory General Purpose RedRod Refillable Glass pH Electrode with Calibration and Maintenance Reagents PackThe RedRod sensor kit comes with calibration buffers reagents and maintenance solutions to provide an all-in-one easy to use reliable solution for your calibrations helping you ensure accurate pH readings with your HQD pH Meter The 500 mL bottles allows up to 6 months of pH electrode calibration when calibrating once per week or up to 25 calibrations at a very attractive price compared if purchased separately
Catalog No. 50-002-07315 Supplier Hach Company Supplier No. PHC70501AP
IntelliCAL PHC705 Laboratory General Purpose RedRod Refillable Glass pH Electrode with Calibration and Maintenance Reagents PackThe RedRod sensor kit comes with calibration buffers reagents and maintenance solutions to provide an all-in-one easy to use reliable solution for your calibrations helping you ensure accurate pH readings with your HQD pH Meter The 500 mL bottles allows up to 6 months of pH electrode calibration when calibrating once per week or up to 25 calibrations at a very attractive price compared if purchased separately