Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Glass Photochemical Reaction Vessel
GREENER_CHOICESDP

Catalog No. 31502355 Shop All Kemtech Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
31-502-355 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31-502-355 Supplier Synthware™ Supplier No. V150500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Internally threaded photochemcial reaction vessel is constructed of borosilicate glass to accomodate an immersion well. The #50 internal thread at the top accepts RC50 bushing and a Viton™ O-ring to form a compression seal with the immersion well.

  • Photochemical reaction vessel constructed of borosilicate glass
  • Internal thread at the top accepts a RC50 bushing and Viton™ O-ring to form a compression seal with the immersion well
  • Includes one 14/20 joint for a sparger tube, one 24/40 vertical joint for a condenser, and one #7 internal thread side arm for a thermometer
  • Complete with quartz immersion well, caps, and O-rings

Specifications

Color Clear
Capacity (Metric) 500 mL
Material Borosilicate Glass (3.3)
For Use With (Application) Low Temperature Reactions
For Use With (Equipment) RC50 Bushing, Viton™ O-Ring
Product Type Reaction Vessel
Side Arm Openings #7
Socket Joint 14/20, 24/40
Thread Size #50
Autoclavable Not Autoclavable
Disposable Reusable
Graduated No
Mouth Style Joint
Sterility Non-sterile
Bottom Shape Round
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.