Learn More
World Precision Instrument Piggyback Glass Capillaries-PB150F-6
SDP
Catalog No. 50821925
Encompass
Piggyback glass capillaries consist of a pair of borosilicate capillaries fused together during manufacture. One barrel is larger than the other, and both have inner filaments for quick filling. Piggyback glass makes it simple to fabricate two-barrel electrodes with a significant tip diameter differential.
Catalog No. 50-821-925 Supplier World Precision Instrument Supplier No. PB150F6
Piggyback glass capillaries consist of a pair of borosilicate capillaries fused together during manufacture. One barrel is larger than the other, and both have inner filaments for quick filling. Piggyback glass makes it simple to fabricate two-barrel electrodes with a significant tip diameter differential.