Learn More
Bellco Glass, Inc. Glass Probe Chamber Assembly , 12mm
SDP
Catalog No. 501946385
Encompass_Preferred
Glass Probe Chamber. MADE IN THE USA-- Glass chamber with hose connections andcompression fitting for 12mm OD probes.Used with any 12mm OD probes – to measure parameters fromprocess such as an off line pH.--Type 1 Borosilicate Glass Chamber--Two Hose Barb Connections--Replacement Cap: A12-1123--Replacement Gasket: A12-1124--Dimensions Approx. 35mm X 25mm OD--Cap Material Hi-Temperature (PBT)
Catalog No. 50-194-6385 Supplier Bellco Glass, Inc. Supplier No. 780800012
Glass Probe Chamber. MADE IN THE USA-- Glass chamber with hose connections andcompression fitting for 12mm OD probes.Used with any 12mm OD probes – to measure parameters fromprocess such as an off line pH.--Type 1 Borosilicate Glass Chamber--Two Hose Barb Connections--Replacement Cap: A12-1123--Replacement Gasket: A12-1124--Dimensions Approx. 35mm X 25mm OD--Cap Material Hi-Temperature (PBT)