Learn More
Hach Company GLASS ROD SECONDARY STD 0.1
Supplier:  Hach Company LZY901
Encompass
  • Provides accurate turbidity reference
  • Makes routine instrument checks easier
  • Eliminates need for liquid calibration standards
  • Compatible with TU5200, TU5300sc, TU5400sc Laser Turbidimeters
  • Includes RFID for easy tracking
  • Enhances workflow efficiency
  • Reduces potential for error in measurements
  • Ideal for routine checks and confirmation of measurements
  • Physical form: Glass rod
  • Turbidity value: Less than 0.1 NTU
Catalog No. NC1998431