Learn More
Microsolv Technology Corporation Glass Rsa Vial/Blue Cap 100
Catalog No. NC1767111
Encompass_Preferred
Vial & Cap Kit Includes 100 EA 1.2ml, MRQ, Screw Top, Clear RSA™ Autosampler Vials and 100 EA matching Screw Caps with White Silicone Rubber / Red PTFE Soft-Guard Septa, bonded in the Blue Caps. RSA Brand Easy Purchase Pack. 1/PK.
Catalog No. NC1767111 Supplier Microsolv Technology Corporation Supplier No. 9512S1EMPRS
Vial & Cap Kit Includes 100 EA 1.2ml, MRQ, Screw Top, Clear RSA™ Autosampler Vials and 100 EA matching Screw Caps with White Silicone Rubber / Red PTFE Soft-Guard Septa, bonded in the Blue Caps. RSA Brand Easy Purchase Pack. 1/PK.