Learn More
Electron Microscopy Sciences Glass Sample Bottle With Plastic Snap Cap, 12 ml
SDP
Catalog No. 5026143
Encompass_Preferred
For collecting and exhibiting a wide variety of specimens in the lab or field. Caps snap over a special sealing ring to form an effective barrier to moisture, making these bottles especially suitable for hydroscopic materials.
Capacity: 12 ml
Catalog No. 50-261-43 Supplier Electron Microscopy Sciences Supplier No. 64254
For collecting and exhibiting a wide variety of specimens in the lab or field. Caps snap over a special sealing ring to form an effective barrier to moisture, making these bottles especially suitable for hydroscopic materials.
Capacity: 12 ml