Learn More
Electron Microscopy Sciences Stereo Viewer Glass Scale with 200 mm Marking Area, 220 x 25 x 3 mm
SDP
Catalog No. 5026147
Encompass_Preferred
A transparent glass plate well polished and marked by the photo-printing technique. It provides very accurate readings and it is a helpful tool for the stereo viewer. Thickness: 3mm.
Marking Area: 200 mm
Size: 220 x 25 x 3 mm
Increment: 0.1 mm
Catalog No. 50-261-47 Supplier Electron Microscopy Sciences Supplier No. 68142
A transparent glass plate well polished and marked by the photo-printing technique. It provides very accurate readings and it is a helpful tool for the stereo viewer. Thickness: 3mm.
Marking Area: 200 mm
Size: 220 x 25 x 3 mm
Increment: 0.1 mm