Learn More
Foxx Life Sciences Borosil Squibb Pear-Shaped Glass Separatory Funnels with Glass Stoppers and PTFE Stopcocks, 500mL, 10/CS
Catalog No. 501542878
Encompass_Preferred
Borosil Separatory Funnels use a self-lubricating, replaceable PTFE stopcock that does not require grease or lubrication in order to achieve fine, smooth control of flow. Glass pennyhead stoppers included. For the best performance in the world, check out Borosil Boroflo Screw-Thread Separatory Funnels (Product Family 6402). Borosil Separatory Funnels comply with DIN/ISO 4800. 60mL & 125mL options are not covered in DIN/ISO. Avail. from 60mL to 2L. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil Type 3.3 Borosilicate glassware to North America for the first time. Manuf. from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity & reliability at competitive prices. And thanks to a unique annealing process, Borosil Glassware is stress-free & incredibly resistant to thermal shock.
Catalog No. 50-154-2878 Supplier Foxx Life Sciences Supplier No. 6403024
Add to Cart
Add to Cart
Borosil Separatory Funnels use a self-lubricating, replaceable PTFE stopcock that does not require grease or lubrication in order to achieve fine, smooth control of flow. Glass pennyhead stoppers included. For the best performance in the world, check out Borosil Boroflo Screw-Thread Separatory Funnels (Product Family 6402). Borosil Separatory Funnels comply with DIN/ISO 4800. 60mL & 125mL options are not covered in DIN/ISO. Avail. from 60mL to 2L. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil Type 3.3 Borosilicate glassware to North America for the first time. Manuf. from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity & reliability at competitive prices. And thanks to a unique annealing process, Borosil Glassware is stress-free & incredibly resistant to thermal shock.