Learn More
Bellco Glass, Inc. Glass Shaft,3L 238mm
SDP
Catalog No. 501945162
Encompass_Preferred
Type 1 A Borosilicate glass rod impeller shaft for 3-liter flask. Glass Shaft,3L238mm. MADE IN THE USA
Catalog No. 50-194-5162 Supplier Bellco Glass, Inc. Supplier No. 196970238
Type 1 A Borosilicate glass rod impeller shaft for 3-liter flask. Glass Shaft,3L238mm. MADE IN THE USA