Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ace Glass, Inc. Glass sheath, 5mm ID, 12.5in, use with 12103-21 probe
SDP

Catalog No. 50977450
Encompass

Glass sheath, 5mm ID, 12.5in, use with 12103-21 probe

Catalog No. 50-977-450 Supplier Ace Glass, Inc. Supplier No. 1210322
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glass sheath, 5mm ID, 12.5in, use with 12103-21 probe

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.