Learn More
Research Products International Corp Glass Shell Vial, 1 Dram, 144 per Case
SDP
Catalog No. 50998595
Encompass
General purpose vial accommodates a variety of laboratory applications. Manufactured from high quality borosilicate glass. Overall vial dimensions are 15 mm x 45 mm with a maximum capacity of 5ml. Supplied with a tight fitting polyethylene stopper not attached to vial. Packaged in a sturdy shelf pack containing 144 vials and stoppers.
Catalog No. 50-998-595 Supplier Research Products International Corp Supplier No. 609650
General purpose vial accommodates a variety of laboratory applications. Manufactured from high quality borosilicate glass. Overall vial dimensions are 15 mm x 45 mm with a maximum capacity of 5ml. Supplied with a tight fitting polyethylene stopper not attached to vial. Packaged in a sturdy shelf pack containing 144 vials and stoppers.