Learn More
Med Vet International Miltex Slab Glass, 4in X 4.5in X .25in, Each, 558-25871
Catalog No. 502831282
Encompass_Preferred
Glass mixing slabs have beveled edges. They are used for mixing cements and paste materials. Teflon coated to prevent material from sticking on the glass. 4in X 4.5in X .25in.
Catalog No. 50-283-1282 Supplier Med Vet International Supplier No. MIL55825871
Glass mixing slabs have beveled edges. They are used for mixing cements and paste materials. Teflon coated to prevent material from sticking on the glass. 4in X 4.5in X .25in.