Learn More
Med Vet International Miltex Slab Glass, Frosted, 4in X 2in X .25in, Each, 558-25875
Catalog No. 502832272
Encompass_Preferred
Glass mixing slabs have beveled edges. They are used for mixing cements and paste materials. Teflon coated to prevent material from sticking on the glass. Frosted. 4in X 2in X .25in.
Catalog No. 50-283-2272 Supplier Med Vet International Supplier No. MIL55825875
Glass mixing slabs have beveled edges. They are used for mixing cements and paste materials. Teflon coated to prevent material from sticking on the glass. Frosted. 4in X 2in X .25in.