Learn More
Electron Microscopy Sciences Glass Slide Frosted End
SDP
Supplier:  Electron Microscopy Sciences 7187001
Encompass_Preferred
The microscope slides have 3/4" sandblasted frosting on one end which allows for easy marking. The slides are made from the highest quality clear-white glass sheets that is fully annealed to minimize chipping. Our microscope glass slides are precleaned and are individually inspected and free from moisture, dirt, and film.
Catalog No. 50-365-976