Learn More
Electron Microscopy Sciences Microscope Slide Label SLRP-15, Pathology, 15/16" x 15/16"
SDP
Catalog No. 5029961
Encompass_Preferred
These specialty Microscope Slide Labels and matching End Labels are available in standard (thin) or pathology (tissue high) thickness, and square or round corner (RC). Permanent adhesive holds labels in place during use and long-term storage. Sheet Form Size is 5 1/4" x 8". Pathology tissue high thickness. Square corners.
Use: Pathology
Size: 15/16" x 15/16"
Catalog No. 50-299-61 Supplier Electron Microscopy Sciences Supplier No. 7702010
These specialty Microscope Slide Labels and matching End Labels are available in standard (thin) or pathology (tissue high) thickness, and square or round corner (RC). Permanent adhesive holds labels in place during use and long-term storage. Sheet Form Size is 5 1/4" x 8". Pathology tissue high thickness. Square corners.
Use: Pathology
Size: 15/16" x 15/16"