Learn More
Electron Microscopy Sciences Glass Slide Plain, 25x75, 1.2 mm thick
SDP
Supplier:  Electron Microscopy Sciences 7188020
Encompass_Preferred
Our microscope glass slides are made from the highest quality clear-white glass sheets and have dimensions that have been standardized to millimeters according to the U.S. government specifications. The glass is fully annealed to minimize chipping.
Our microscope glass slides are precleaned and are individually inspected and free from moisture, dirt, and film. All of our slides are flat, uniform and durable.
Size: 25x75
Thickness: 1.2 mm
Catalog No. 50-949-789