Learn More
Electron Microscopy Sciences Glass Slide Processing Holder
SDP
Catalog No. 50931161
Encompass_Preferred
The holder is designed to hold securely five 1x3" microscope slides. It comes with a handle for easy manipulation. Up to 50 slides (10 holders) can be processed at one time in one staining vessel from start to finish without interruption.
The slide holder has two sides-one being black and the other white. This allows for easy grouping and identification of the slides. The holder is ideal for both frozen and paraffin sections.
Slide holder can be used to fix frozen sections in acetone, deparaffinize and rehydrate paraffin sections, isolate sections on the slide with the PAP pen, immunostain, incubate and wash, and counter stain, mounting with medium.
Catalog No. 50-931-161 Supplier Electron Microscopy Sciences Supplier No. 7152401
The holder is designed to hold securely five 1x3" microscope slides. It comes with a handle for easy manipulation. Up to 50 slides (10 holders) can be processed at one time in one staining vessel from start to finish without interruption.
The slide holder has two sides-one being black and the other white. This allows for easy grouping and identification of the slides. The holder is ideal for both frozen and paraffin sections.
Slide holder can be used to fix frozen sections in acetone, deparaffinize and rehydrate paraffin sections, isolate sections on the slide with the PAP pen, immunostain, incubate and wash, and counter stain, mounting with medium.