Learn More
Bruker Daltonics MALDI IntelliSlides, Box of 50 slides, individually barcoded
Catalog No. NC0380464
Encompass
Bruker’s IntelliSlides is a dedicated solution designed to simplify your MALDI Imaging workflows by enabling automated setup of each measurement. IntelliSlides incorporate permanent inscriptions that indicates optimal placement of samples, registration marks (also known as teach marks or fiducials) that make automated sample registration possible, as well as providing a unique slide identifier for software tracking. IntelliSlides are the optimal tool for increasing measurement efficiency and throughput for any MALDI Imaging study
Catalog No. NC0380464 Supplier Bruker Daltonics Supplier No. 1868957
Bruker’s IntelliSlides is a dedicated solution designed to simplify your MALDI Imaging workflows by enabling automated setup of each measurement. IntelliSlides incorporate permanent inscriptions that indicates optimal placement of samples, registration marks (also known as teach marks or fiducials) that make automated sample registration possible, as well as providing a unique slide identifier for software tracking. IntelliSlides are the optimal tool for increasing measurement efficiency and throughput for any MALDI Imaging study