Learn More
Sigma Aldrich Fine Chemicals Biosciences Glass spreaders 90 bend
Catalog No. 502900671
Encompass
Constructed of 4 mm diameter glass rod with polished ends with 130 mm long handles and 50 mm long spreader segments.
Catalog No. 50-290-0671 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S45226EA
Constructed of 4 mm diameter glass rod with polished ends with 130 mm long handles and 50 mm long spreader segments.