Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Glass Square Media Bottles

Catalog No. 22443938
Change view
Click to view available options
Capacity (Metric):
500 mL
1 L
2 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
22-443-938 1 L
22-443-950 500 mL
22-443-939 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-443-938 Supplier Globe Scientific Supplier No. 8121000
Add to Cart
Add to Cart

Globe Glass™ square wide mouth media bottles are designed with an extra wide mouth for ease of filling, dispensing, and pipetting.

  • The square shape improves storage and transport efficiency.
  • A drip free pour ring and autoclavable polypropylene (PP) screw cap enable sterilization of media and reagents.
  • Globe Glass™ square wide mouth media bottles are available in sizes 500 mL, 1000 mL, and 2000 mL.
  • ASTM E438, Type I, Class A, Borosilicate 3.3 Glass
  • Square shape improves storage efficiency
  • GlassGuard™ packaging system protects against transit damage
  • GL80 polypropylene (PP) screw caps
  • Pouring ring for drip free liquid transfer
  • High resistance to chemicals
  • Single scale metric graduations
  • Durable white enamel printing
  • Large white marking area

Specifications

Capacity (Metric) 1 L
Capacity (English) 33.81 oz.
Color Clear
Material Borosilicate Glass (3.3)
Sterility Non-sterile
Closure Included Yes
Disposable Reusable
Graduated Yes
Mouth Threaded
Shape Square
Product Type Media Storage Bottle
Closure Size GL80
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.