Learn More
Description
- The square shape improves storage and transport efficiency.
- A drip free pour ring and autoclavable polypropylene (PP) screw cap enable sterilization of media and reagents.
- Globe Glass™ square wide mouth media bottles are available in sizes 500 mL, 1000 mL, and 2000 mL.
- ASTM E438, Type I, Class A, Borosilicate 3.3 Glass
- Square shape improves storage efficiency
- GlassGuard™ packaging system protects against transit damage
- GL80 polypropylene (PP) screw caps
- Pouring ring for drip free liquid transfer
- High resistance to chemicals
- Single scale metric graduations
- Durable white enamel printing
- Large white marking area
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Clear |
| Material | Borosilicate Glass (3.3) |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | Yes |
| Mouth | Threaded |
| Closure Size | GL80 |
| Autoclavable | Autoclavable |
| Capacity (English) | 16.91 oz. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.