Learn More
Newcomer Supply 20 Slide Staining Dish, Glass (Includes Lid with Gasket Seal)
SDP
Catalog No. NC2102243
Encompass
The Glass Staining Dish accommodates a 20 slide rack and is made of a heavy glass. The lid has a snug fit with a gasket seal. Inside Dimensions: 3 3/4" x 3" x 2 1/4" (H), Outside Dimensions: 4 1/4" x 3 5/8" x 3" (H). Dishes are also available in 2 other sizes to accommodate 30 and 60 place slide racks.
Catalog No. NC2102243 Supplier Newcomer Supply Supplier No. 6816A
The Glass Staining Dish accommodates a 20 slide rack and is made of a heavy glass. The lid has a snug fit with a gasket seal. Inside Dimensions: 3 3/4" x 3" x 2 1/4" (H), Outside Dimensions: 4 1/4" x 3 5/8" x 3" (H). Dishes are also available in 2 other sizes to accommodate 30 and 60 place slide racks.