Learn More
Research Products International Corp Glass Staining Dish, Small, 22 x 33 cm
SDP
Catalog No. 50998468
Encompass
These glass dishes are ideal for staining, destaining and fixing applications using large format blots. Dishes are molded from clear Pyrex borosilicate glass for durability and ease of cleaning. Dishes are 5cm deep and feature carrying handles for convenient handling. Safe for microwave use.
Catalog No. 50-998-468 Supplier Research Products International Corp Supplier No. 248932
These glass dishes are ideal for staining, destaining and fixing applications using large format blots. Dishes are molded from clear Pyrex borosilicate glass for durability and ease of cleaning. Dishes are 5cm deep and feature carrying handles for convenient handling. Safe for microwave use.