Learn More
Newcomer Supply Staining Dish for 30 Slide Rack
SDP
Catalog No. NC0779539
Encompass
The Glass Staining Dish accommodates a 30 slide rack and is made of a heavy glass. The lid has a snug fit with a gasket seal. Inside Dimensions: 3 3/4" x 3 1/4" x 3 3/8" (H), Outside Dimensions: 4" x 4 1/8" x 4" (H). Dishes are also available in 2 other sizes to accommodate 20 and 60 place slide racks.
Catalog No. NC0779539 Supplier Newcomer Supply Supplier No. 6816B
The Glass Staining Dish accommodates a 30 slide rack and is made of a heavy glass. The lid has a snug fit with a gasket seal. Inside Dimensions: 3 3/4" x 3 1/4" x 3 3/8" (H), Outside Dimensions: 4" x 4 1/8" x 4" (H). Dishes are also available in 2 other sizes to accommodate 20 and 60 place slide racks.