Learn More
Newcomer Supply Glass Staining Dish for 60 Slide Rack
SDP
Catalog No. NC1550126
Encompass
The Glass Staining Dish accommodates a 60 slide rack and is made of a heavy glass. The lid has a snug fit with a gasket seal. Inside Dimensions: 6" x 3 1/4" x 3 3/8" (H), Outside Dimensions: 6 3/8" x 3 3/4" x 4" (H). Dishes are also available in 2 other sizes to accommodate 20 and 30 place slide racks.
Catalog No. NC1550126 Supplier Newcomer Supply Supplier No. 6816C
The Glass Staining Dish accommodates a 60 slide rack and is made of a heavy glass. The lid has a snug fit with a gasket seal. Inside Dimensions: 6" x 3 1/4" x 3 3/8" (H), Outside Dimensions: 6 3/8" x 3 3/4" x 4" (H). Dishes are also available in 2 other sizes to accommodate 20 and 30 place slide racks.