Learn More
Cole-Parmer DWK Life Sciences (Wheaton) Glass Coplin Staining Jar with glass cover
Catalog No. NC0089608 Shop All Cole-Parmer Products
Encompass_Preferred
Heavy glass jar provides a stable container for staining slides. Glass cover protects slides from outside elements and reduces reagent evaporation. Requires only a small volume of reagent. Holds up to ten 75 x 25 mm (3'' x 1'') slides back to back. Inside measures 30 mm W x 75 mm H x 30 mm D.
Catalog No. NC0089608 Supplier Cole-Parmer Supplier No. 4858530
Heavy glass jar provides a stable container for staining slides. Glass cover protects slides from outside elements and reduces reagent evaporation. Requires only a small volume of reagent. Holds up to ten 75 x 25 mm (3'' x 1'') slides back to back. Inside measures 30 mm W x 75 mm H x 30 mm D.