Learn More
Research Products International Corp Glass Staining Dish with Lid, Small, 11 x 15 cm, 2 per Package
SDP
Catalog No. 50550322
Encompass
Ideal for all staining, fixing and destaining procedures. Heavy duty staining dishes are molded from clear Pyrex brand borosilicate glass. Offers best compatibility with all chemical buffers and reagents. Safe to use with electrophoresis stains and dyes. Clean thoroughly after use. Supplied with tight fitting polyethylene lids to prevent evaporation of solutions.
Catalog No. 50-550-322 Supplier Research Products International Corp Supplier No. 248924
Ideal for all staining, fixing and destaining procedures. Heavy duty staining dishes are molded from clear Pyrex brand borosilicate glass. Offers best compatibility with all chemical buffers and reagents. Safe to use with electrophoresis stains and dyes. Clean thoroughly after use. Supplied with tight fitting polyethylene lids to prevent evaporation of solutions.