Learn More
Cole-Parmer Hamilton 80300 Standard Microliter Syringes, 10 uL, Cemented-Needle, 1/ea
Catalog No. NC1045883 Shop All Cole-Parmer Products
Encompass_Preferred
Microliter syringes are available in volumes from 10 µL to 500 µL and provide precise liquid delivery. Choose from cemented-needle, removable-needle, or Luer tip syringes (needles must be purchased separately for Luer tip syringes). Needles for these syringes have a 22° bevel; designed for septum penetration in all chromatographic techniques. If you use large numbers of syringes, choose one of our convenient economy packs containing six 10-µL syringes with cemented or removable needles. Each kit is packaged in a protective storage container and includes a set of adhesive-backed numbers to aid in identifying syringes containing different samples. Syringes with luer tip connections do not include a needle, order needles separately.
Catalog No. NC1045883 Supplier Cole-Parmer Supplier No. 0793800
Microliter syringes are available in volumes from 10 µL to 500 µL and provide precise liquid delivery. Choose from cemented-needle, removable-needle, or Luer tip syringes (needles must be purchased separately for Luer tip syringes). Needles for these syringes have a 22° bevel; designed for septum penetration in all chromatographic techniques. If you use large numbers of syringes, choose one of our convenient economy packs containing six 10-µL syringes with cemented or removable needles. Each kit is packaged in a protective storage container and includes a set of adhesive-backed numbers to aid in identifying syringes containing different samples. Syringes with luer tip connections do not include a needle, order needles separately.