Learn More
Cole-Parmer DWK Life Sciences (Kimble) Stir Rod, Glass, 5 mm dia X 150 mm L, 25/pk
Catalog No. NC9747965 Shop All Cole-Parmer Products
Encompass_Preferred
Manufactured with borosilicate glass with rounded annealed ends. Stir rods are highly chemical resistant and withstands temperatures to 400 F (205)C. Autoclavable.
Catalog No. NC9747965 Supplier Cole-Parmer Supplier No. 0636921
Manufactured with borosilicate glass with rounded annealed ends. Stir rods are highly chemical resistant and withstands temperatures to 400 F (205)C. Autoclavable.