Learn More
Cole-Parmer DWK Life Sciences (Kimble) Stir Rod, Glass, 6.4 mm dia X 250 mm L, 25/pk
Catalog No. NC9747966 Shop All Cole-Parmer Products
Encompass_Preferred
Manufactured with borosilicate glass with rounded annealed ends. Stir rods are highly chemical resistant and withstands temperatures to 400° F (205°)C. Autoclavable.
Catalog No. NC9747966 Supplier Cole-Parmer Supplier No. 0636923
Manufactured with borosilicate glass with rounded annealed ends. Stir rods are highly chemical resistant and withstands temperatures to 400° F (205°)C. Autoclavable.