Learn More
Cole-Parmer Cole-Parmer Glass Stir Rods with Rubber Policeman, 8" 12/pk
Catalog No. NC1345381 Shop All Cole-Parmer Products
Encompass_Preferred
Glass stir rods are manufactured with soft borosilicate glass with both ends rounded. They come with a pre-assembled, removable rubber policeman for transferring liquids or clearing precipitate out of containers. Glass rods are chemically resistant and can withstand temperatures to 400°F (205°C) (without rubber policemen). Autoclavable.
Catalog No. NC1345381 Supplier Cole-Parmer Supplier No. 0636954
Glass stir rods are manufactured with soft borosilicate glass with both ends rounded. They come with a pre-assembled, removable rubber policeman for transferring liquids or clearing precipitate out of containers. Glass rods are chemically resistant and can withstand temperatures to 400°F (205°C) (without rubber policemen). Autoclavable.