Learn More
Foxx Life Sciences Borosil® Reusable Solid Stirring Rods, Polished Glass Stirrers, 9mm x 205mm (MediumLength),20/CS
Supplier:  Foxx Life Sciences 9850209
Encompass_Preferred
Borosil® Polished Glass Solid Stirring Rods are constructed from smooth, low-expansion, low-extractable borosilicate glass. Their mechanical strength, excellent heat resistance, and outstanding chemical compatibility make them the perfect general use stirrer. Borosil® Glass Stirrers are avaialble in two widths: 7mm and Extra-Wide 9mm. Each width is available in four lengths, from the Short 150mm to the Extra Large 305mm. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-172-0213