Learn More
Electron Microscopy Sciences EMS Glass Knife Strip Ultramicrotomy Grade, 10 mm thick, 25.4 x 203 mm
SDP
Supplier:  Electron Microscopy Sciences 12356
Encompass_Preferred
Specially selected glass strips for making glass knives. Its hardness produces a durable and sharp edge suitable for thick, semi thin, or ultrathin sections.
Thickness: 10 mm
Size: 25.4 x 203 mm
Catalog No. 50-193-0513