Learn More
Electron Microscopy Sciences EMS Glass Knife Strip Ultramicrotomy Grade, 10 mm thick, 50.8 x 101 mm
SDP
Catalog No. 501930515
Encompass_Preferred
Specially selected glass strips for making glass knives. Its hardness produces a durable and sharp edge suitable for thick, semi thin, or ultrathin sections.
Thickness: 10 mm
Size: 50.8 x 101 mm
Catalog No. 50-193-0515 Supplier Electron Microscopy Sciences Supplier No. 12358
Specially selected glass strips for making glass knives. Its hardness produces a durable and sharp edge suitable for thick, semi thin, or ultrathin sections.
Thickness: 10 mm
Size: 50.8 x 101 mm