Learn More
Grainger Reusable Glass Syringe, Glass Body Material, 5 mL Capacity
Catalog No. NC1570307
Encompass_Preferred
High-purity borosilicate glass syringe is reusable for laboratory and research applications. Barrel and piston are interchangeable. CE Certified. Individually boxed.
Catalog No. NC1570307 Supplier Grainger Supplier No. 19G360
High-purity borosilicate glass syringe is reusable for laboratory and research applications. Barrel and piston are interchangeable. CE Certified. Individually boxed.