Learn More
Cole-Parmer Glass Tempering Beaker, borosilicate, 1/2" hose barbs, 250 mL
Catalog No. NC1661054 Shop All Cole-Parmer Products
Encompass_Preferred
Liquid-jacketed borosilicate glass vessels have double-walled casing for circulating cold or warm tempering fluid. Tubing connectors measure 1 1⁄2"L x 1⁄2" OD. Connect a constant temperature circulator to maintain sample temperatures.
Catalog No. NC1661054 Supplier Cole-Parmer Supplier No. 0377310
Liquid-jacketed borosilicate glass vessels have double-walled casing for circulating cold or warm tempering fluid. Tubing connectors measure 1 1⁄2"L x 1⁄2" OD. Connect a constant temperature circulator to maintain sample temperatures.