Learn More
Cole-Parmer Glass Tempering Beaker, borosilicate, 1/2" hose barbs, 2000 mL
Catalog No. NC1430212 Shop All Cole-Parmer Products
Encompass_Preferred
Liquid-jacketed borosilicate glass vessels have double-walled casing for circulating cold or warm tempering fluid. Tubing connectors measure 1 1⁄2"L x 1⁄2" OD. Connect a constant temperature circulator to maintain sample temperatures.
Catalog No. NC1430212 Supplier Cole-Parmer Supplier No. 0377330
Add to Cart
Add to Cart
Liquid-jacketed borosilicate glass vessels have double-walled casing for circulating cold or warm tempering fluid. Tubing connectors measure 1 1⁄2"L x 1⁄2" OD. Connect a constant temperature circulator to maintain sample temperatures.