Learn More
World Precision Instrument Septum Theta
SDP
Catalog No. 50821979
Encompass
Quality glass, superior prices for microinjection/microelectrodes
Superior Cell Impalement
Septum Theta offers superior cell impalement. The natural bevel resulting from the prominent spear-like projection of the septum gives microelectrodes a sharp, spear-point tip. This style has low resistance for use as a single microelectrode, and it can be used to make superior double-tipped microelectrodes with low trans-tip coupling. The natural bevel of Septum Theta also significantly increases the effective tip cross-section. As supplied, the width of the septum is approximately 0.2 mm, and the wall thickness is approximately 0.2 mm.
Catalog No. 50-821-979 Supplier World Precision Instrument Supplier No. TST1506
Quality glass, superior prices for microinjection/microelectrodes
Superior Cell Impalement
Septum Theta offers superior cell impalement. The natural bevel resulting from the prominent spear-like projection of the septum gives microelectrodes a sharp, spear-point tip. This style has low resistance for use as a single microelectrode, and it can be used to make superior double-tipped microelectrodes with low trans-tip coupling. The natural bevel of Septum Theta also significantly increases the effective tip cross-section. As supplied, the width of the septum is approximately 0.2 mm, and the wall thickness is approximately 0.2 mm.