Learn More
Miles Scientific Latch-Lid Developing Chamber for 20x20 cm TLC Plates
SDP
Catalog No. NC9014009
Encompass_Preferred
Analtech's glass TLC developing chambers provide optimum developing conditions for a variety of TLC plate sizes and shapes. Precision ground surfaces provide tight seals between tanks and glass covers and permit complete saturation of chambers with solvent vapor for uniform development of chromatograms. Each chamber is shipped with the appropriate lid. Inside Dimensions: 25 cm H x 28 cm W x 7.5 cm D. For use with: 20 x 20 cm TLC plates or smaller.
Catalog No. NC9014009 Supplier Miles Scientific Supplier No. A8033
Analtech's glass TLC developing chambers provide optimum developing conditions for a variety of TLC plate sizes and shapes. Precision ground surfaces provide tight seals between tanks and glass covers and permit complete saturation of chambers with solvent vapor for uniform development of chromatograms. Each chamber is shipped with the appropriate lid. Inside Dimensions: 25 cm H x 28 cm W x 7.5 cm D. For use with: 20 x 20 cm TLC plates or smaller.