Learn More
Cole-Parmer Griffin Type Glass Tubing Cutter for up to 1-1/2" (38 mm) Diameter, 1/Pk
Catalog No. NC9996565 Shop All Cole-Parmer Products
Encompass_Preferred
Spring loaded cutter cuts glass up to 38mm. Simply insert tubing into the cutter and rotate tube to make a precise, clean cut. Manufactured of nickel-plated zinc alloy, cutter is corrosion resistant.
Catalog No. NC9996565 Supplier Cole-Parmer Supplier No. 3474280
Add to Cart
Add to Cart
Spring loaded cutter cuts glass up to 38mm. Simply insert tubing into the cutter and rotate tube to make a precise, clean cut. Manufactured of nickel-plated zinc alloy, cutter is corrosion resistant.