Learn More
Research Products International Corp Low Background Glass Scintillation Vials, 20ml Capacity, 22mm Cap Size, Urea Unattched Caps, Polyseal Cone Liners, 500 per Case
SDP
Catalog No. 50212994
Encompass
Manufactured from select low background borosilicate glass. Special attention given to all critical dimensions such as wall thickness, vial diameter and length. Compatible with all liquid scintillation counters that accommodate 20ml size vials.
Catalog No. 50-212-994 Supplier Research Products International Corp Supplier No. 121002
Manufactured from select low background borosilicate glass. Special attention given to all critical dimensions such as wall thickness, vial diameter and length. Compatible with all liquid scintillation counters that accommodate 20ml size vials.