Learn More
Research Products International Corp Low Background Glass Scintillation Vials, 20ml Capacity, 22mm Cap Size, Polypropylene Attached Caps, Metal Foil Liners, 500 per Case
SDP
Catalog No. 50476612
Encompass
Manufactured from select low background borosilicate glass. Special attention given to all critical dimensions such as wall thickness, vial diameter and length. Compatible with all liquid scintillation counters that accommodate 20ml size vials.
Catalog No. 50-476-612 Supplier Research Products International Corp Supplier No. 121000CA
Manufactured from select low background borosilicate glass. Special attention given to all critical dimensions such as wall thickness, vial diameter and length. Compatible with all liquid scintillation counters that accommodate 20ml size vials.