Learn More
TWD Scientific LLC Small Glass Weighing Funnel. Pack of 6
SDP
Catalog No. NC9087548 Shop All TWD Scientific LLC Products
Encompass_Preferred
Disposable borosilicate glass weighing funnel, size small. Fits volumetric flask volumes (mL) of 5, 10, 15, and 20. Funnels have a maximum capacity of 2mL Solid/2mL Liquid. 6/Pack
Catalog No. NC9087548 Supplier TWD Scientific LLC Supplier No. GWFBS1S
Disposable borosilicate glass weighing funnel, size small. Fits volumetric flask volumes (mL) of 5, 10, 15, and 20. Funnels have a maximum capacity of 2mL Solid/2mL Liquid. 6/Pack