Learn More
Anton Paar 15.5 mm Glass Window for Polorimeter Cells
Catalog No. NC1410391
Encompass_Preferred
This item has a minimum qty of 5 per supplier requirements.
15.5 mm Glass Window for 50 - 200 mm polarimeter sample cells
Catalog No. NC1410391 Supplier Anton Paar Supplier No. 49703
This item has a minimum qty of 5 per supplier requirements.
15.5 mm Glass Window for 50 - 200 mm polarimeter sample cells