Learn More
Hach Company Glass Wool
Supplier:  Hach Company 58901
Encompass
Pliable and easy to use. Contains no heavy metals. Low alkali content. Suitable for filtering. Comes in sheets 3 mm (0.1") thick x 250 mm (9.8") wide x 1300 mm (51.2") long.
Catalog No. 50-199-4403