Learn More
Crescent Chemical Co Inc GLASS WOOL SILANIZED
SDP
Catalog No. 50247630
Encompass_Preferred
Glass wool, silanized research grade A very soft material for plugging columns in gas and liquid chromatography. Size - 50G Storage Conditions - +15 °C TO +30 °C Catalog Number - 22367.03
Catalog No. 50-247-630 Supplier Crescent Chemical Co Inc Supplier No. SE22367.03
Glass wool, silanized research grade A very soft material for plugging columns in gas and liquid chromatography. Size - 50G Storage Conditions - +15 °C TO +30 °C Catalog Number - 22367.03