Learn More
Crescent Chemical Co Inc GLASS WOOL SILANIZED
SDP
Supplier:  Crescent Chemical Co Inc SE22367.01
Encompass_Preferred
Glass wool, silanized research grade A very soft material for plugging columns in gas and liquid chromatography. Size - 10G Storage Conditions - +15 °C TO +30 °C Catalog Number - 22367.01
Catalog No. 50-247-629