Learn More
Cycle-Tap Sampler, J.T. Baker™
Catalog No. 02003408 Shop All Avantor Products
Change view
Click to view available options
:
DCM, High Purity, Low Water, Green QC
Catalog No.
02-003-408 DCM, High Purity, Low Water, Green QC
1 options
This item is not returnable. View return policy
Catalog No. 02-003-408 Supplier Avantor Supplier No. 981504
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
The CYCLE-TAP sampler is a manually operated device for removing small volumes of solvent from the CYCLETAINER solvent delivery system using a syringe.
Specifications
Chemical Name or Material Cycle-Tap Sampler