Learn More
Grainger Radians Rad-Atac Safety Glasses, Clear, Polycarbonate Lens, 1 ea
Catalog No. 502786825
Encompass_Preferred
These wraparound safety glasses are designed for reliable eye protection in indoor or outdoor environments. They feature a lightweight, single-lens construction that ensures a wide field of vision, combined with an integrated nosepiece and rubber-tipped temples for a secure, comfortable fit throughout the day.
  • Single polycarbonate lens
  • Meets Z87.1 high impact standards
  • Blocks 99.9% of harmful UV rays
  • Soft, non-slip rubber tipped temples
  • Integrated nosepiece
  • Clear lens color
Catalog No. 50-278-6825 Supplier Grainger Supplier No. 198DY8
These wraparound safety glasses are designed for reliable eye protection in indoor or outdoor environments. They feature a lightweight, single-lens construction that ensures a wide field of vision, combined with an integrated nosepiece and rubber-tipped temples for a secure, comfortable fit throughout the day.
  • Single polycarbonate lens
  • Meets Z87.1 high impact standards
  • Blocks 99.9% of harmful UV rays
  • Soft, non-slip rubber tipped temples
  • Integrated nosepiece
  • Clear lens color